Learn More
MSC Detecto AC Adapter, 12 VDC, 1A, 1 ea
Catalog No. 5000795127
Encompass
This AC adapter provides 12 VDC 1A output, serving as an optional power source for various Detecto digital scales and stadiometers, and is included with some -AC models. It is also capable of recharging Ni-Cad or NiMH batteries in compatible weight indicators.
  • Delivers 12 VDC at 1A output
  • Input voltage range of 100–240 VAC
  • UL/CSA listed for safety standards
  • Compatible with various Detecto digital scales and stadiometers, including APEX, 6475, 68XX, IBFL500, 8432-CH, 8435, VET70, DR400-750, AP, APS, RP, EB, and SONARIS models
  • Supports recharging of Ni-Cad or NiMH batteries (do not use with Alkaline batteries)
Catalog No. 50-007-95127 Supplier MSC Supplier No. 65360547
This AC adapter provides 12 VDC 1A output, serving as an optional power source for various Detecto digital scales and stadiometers, and is included with some -AC models. It is also capable of recharging Ni-Cad or NiMH batteries in compatible weight indicators.
  • Delivers 12 VDC at 1A output
  • Input voltage range of 100–240 VAC
  • UL/CSA listed for safety standards
  • Compatible with various Detecto digital scales and stadiometers, including APEX, 6475, 68XX, IBFL500, 8432-CH, 8435, VET70, DR400-750, AP, APS, RP, EB, and SONARIS models
  • Supports recharging of Ni-Cad or NiMH batteries (do not use with Alkaline batteries)