Learn More
MSC Detecto Dial Hanging Scale, 40 lb., 8in Dial, 1 ea
Catalog No. 5000793040
Encompass
The Detecto Dial Hanging Scale is a precision instrument designed for accurate weight measurement, featuring a stainless steel round pan, an 8-in dial display, and a capacity of 40 lb. It is suitable for various weighing applications requiring clear and reliable readings.
  • Features a 40 lb. capacity with 1 oz graduation
  • Has an 8-in dial display with 2 revolutions
  • Constructed with plastic housing, steel hook and frame
  • Equipped with a stainless steel round pan
Catalog No. 50-007-93040 Supplier MSC Supplier No. 65360802
The Detecto Dial Hanging Scale is a precision instrument designed for accurate weight measurement, featuring a stainless steel round pan, an 8-in dial display, and a capacity of 40 lb. It is suitable for various weighing applications requiring clear and reliable readings.
  • Features a 40 lb. capacity with 1 oz graduation
  • Has an 8-in dial display with 2 revolutions
  • Constructed with plastic housing, steel hook and frame
  • Equipped with a stainless steel round pan