Detecto portable weighbeam scales are designed for industrial use, offering precise measurements in various demanding environments. Built with heavy-duty welded steel, these scales provide reliable and maintenance-free operation, making them suitable for intrinsically-safe and hazardous locations where no external power is required.
- Heavy-duty welded steel construction with electrostatic powder paint coating
- NTEP and Measurement Canada certified
- Platform dimensions 19 in W x 28 in D (48 cm W x 71 cm D)
- No power required for complete portability
- Four 5-in (13-cm) diameter rubolene wheels for maneuverability
- Maintenance-free lever system operation
- Die-cast beam with easy-to-read numbers, viewable from front and back
- Suitable for hazardous locations and intrinsically safe environments
- Lightweight design at 116 lb (53 kg) net weight