Learn More
Medchemexpress LLC Deterenol hydrochloride | 23239-36-3 | 99.7% | 231.72 g/mol | C11H18ClNO2 | 5 MG
SDP
Catalog No. 5000284706
Encompass_Preferred
Deterenol hydrochloride is a selective β-adrenergic receptor agonist supplied for research use. It is reported to cause vasodepressor effects, anxiety, increased heart rate, hypotension, palpitations, and respiratory distress; handle with appropriate laboratory safety practices.
  • Selective β-adrenergic agonist for pharmacology research.
  • Reported physiological effects include vasodepression, tachycardia, hypotension, and anxiety.
  • High stated purity: 99.7%.
  • Off-white to light yellow solid suitable for small-scale assays.
  • Typically supplied in milligram quantities and stored sealed at -20°C.
Catalog No. 50-002-84706 Supplier Medchemexpress LLC Supplier No. HYW7020275MG
Deterenol hydrochloride is a selective β-adrenergic receptor agonist supplied for research use. It is reported to cause vasodepressor effects, anxiety, increased heart rate, hypotension, palpitations, and respiratory distress; handle with appropriate laboratory safety practices.
  • Selective β-adrenergic agonist for pharmacology research.
  • Reported physiological effects include vasodepression, tachycardia, hypotension, and anxiety.
  • High stated purity: 99.7%.
  • Off-white to light yellow solid suitable for small-scale assays.
  • Typically supplied in milligram quantities and stored sealed at -20°C.