Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deterenol hydrochloride | 23239-36-3 | MFCD28898982 | 99.7% | 231.72 g/mol | C11H18ClNO2 | 25 MG
SDP

Catalog No. 5000284670
Encompass_Preferred

Deterenol hydrochloride is the hydrochloride salt of deterenol, a selective beta-adrenergic agonist used as a research reagent. Provided as a solid with high purity, it is intended for in vitro and in vivo pharmacology studies involving beta-adrenergic receptors and cardiovascular response assessment.

  • Selective beta-adrenergic agonist activity.
  • High purity suitable for research applications.
  • Hydrochloride salt form for improved stability and handling.
  • Common use in receptor pharmacology and cardiovascular studies.
  • Molecular formula C11H18ClNO2; molecular weight 231.72 g/mol.
  • CAS number 23239-36-3 for unambiguous identification.

Catalog No. 50-002-84670 Supplier Medchemexpress LLC Supplier No. HYW70202725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deterenol hydrochloride is the hydrochloride salt of deterenol, a selective beta-adrenergic agonist used as a research reagent. Provided as a solid with high purity, it is intended for in vitro and in vivo pharmacology studies involving beta-adrenergic receptors and cardiovascular response assessment.

  • Selective beta-adrenergic agonist activity.
  • High purity suitable for research applications.
  • Hydrochloride salt form for improved stability and handling.
  • Common use in receptor pharmacology and cardiovascular studies.
  • Molecular formula C11H18ClNO2; molecular weight 231.72 g/mol.
  • CAS number 23239-36-3 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.