Learn More
Crescent Chemical Co Inc DETERGENT 7 X NEUTRAL PHOS
SDP
Supplier:  Crescent Chemical Co Inc SE34205.02
Encompass_Preferred
Detergent 7 X neutral, phosphate-free For tissue culture and molecular biology. Highly active cleaning material, guaranteed to be non-toxic for even the most sensitive organisms. Size - 10L Storage Conditions - +15 C TO +30 C Catalog Number - 342
Catalog No. 50-248-094