Learn More
Active Motif Detergent
SDP
Catalog No. NC1391874
Encompass
Active Motif's non-ionic detergent can be added to wash buffers to help increase stringency.
Catalog No. NC1391874 Supplier Active Motif Supplier No. 37517
Add to Cart
Edge
Add to Cart
Active Motif's non-ionic detergent can be added to wash buffers to help increase stringency.