Learn More
Sigma Aldrich Fine Chemicals Biosciences Detergent 8R liquid deterg1EA
Catalog No. 502901347
Encompass
Low-foaming Ion-free DetergentReplace solvent cleaningConcentrated to save you moneyBiodegradable and readily disposablePenetrating wetting power to save you timefree rinsing to give you reliable results and no interfering residuesContains no phosphate sulfur chlorine metal cation halide borate silicate carbonate chlorocarbon fluorocarbon or chelating ingredientsUsed to clean Circuit boards electronic parts phosphate sensitive labware nuclear reactor cavities delicate industrial parts and nuclear contaminated equipment during outage. fDA certified.Used to remove Resins rosins oils dirt grime particulates residues fluxes chemicals and solvents.Surfaces cleaned Corrosion inhibited formulation recommended for glass metal stainless steel ceramic plastic and circuit boards. Can be used with prompt rinsing on soft metals such as copper aluminum silver and gold. Aluminum can darken and mixed metal galvanic reactions can occur. Corro
Catalog No. 50-290-1347 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2784911EA
Low-foaming Ion-free DetergentReplace solvent cleaningConcentrated to save you moneyBiodegradable and readily disposablePenetrating wetting power to save you timefree rinsing to give you reliable results and no interfering residuesContains no phosphate sulfur chlorine metal cation halide borate silicate carbonate chlorocarbon fluorocarbon or chelating ingredientsUsed to clean Circuit boards electronic parts phosphate sensitive labware nuclear reactor cavities delicate industrial parts and nuclear contaminated equipment during outage. fDA certified.Used to remove Resins rosins oils dirt grime particulates residues fluxes chemicals and solvents.Surfaces cleaned Corrosion inhibited formulation recommended for glass metal stainless steel ceramic plastic and circuit boards. Can be used with prompt rinsing on soft metals such as copper aluminum silver and gold. Aluminum can darken and mixed metal galvanic reactions can occur. Corro