Learn More
Andwin Scientific DETERGENT ALCONOX
SDP
Catalog No. NC9003111
Encompass_Preferred
The Alconox powder detergent is a suitable product for eliminating soil, grit, buffing compounds, and other organic pollutants. This product is compatible to use with glass, metal, stainless steel, porcelain, ceramic, plastic, rubber, and fiberglass. The concentrated detergent is noncorrosive and is made with high-foaming formula.
This item is not returnable. View return policy
Catalog No. NC9003111 Supplier Andwin Scientific Supplier No. 21835
The Alconox powder detergent is a suitable product for eliminating soil, grit, buffing compounds, and other organic pollutants. This product is compatible to use with glass, metal, stainless steel, porcelain, ceramic, plastic, rubber, and fiberglass. The concentrated detergent is noncorrosive and is made with high-foaming formula.