Learn More
Hach Company Detergent-alcotabs
Catalog No. 5000256958 Shop All Hach Company Products
Encompass
Effervescent detergent tablets for maximum cleaning in syphon pipette washers to clean pipettes and tubes Slow release bubble action and free rinsing produce reliable results and no interfering residues pH 6 5 Contains phosphate
Catalog No. 50-002-56958 Supplier Hach Company Supplier No. 2088200
Effervescent detergent tablets for maximum cleaning in syphon pipette washers to clean pipettes and tubes Slow release bubble action and free rinsing produce reliable results and no interfering residues pH 6 5 Contains phosphate