Learn More
Apexbio Technology LLC Detergent Compatible Bradford Protein Assay Kit 800T
SDP
Catalog No. 5000097259
Encompass_Preferred
The Detergent Compatible Bradford Protein Assay Kit is designed for accurate quantification of protein concentrations in samples containing detergents Utilizing the Bradford assay principle the kit employs Coomassie Brilliant Blue dye which binds to proteins primarily through arginine and aromatic residues resulting in a measurable shift in absorbance Unlike traditional Bradford assays this formulation maintains assay performance in the presence of common detergents minimizing interference and ensuring reliable results The kit is suitable for high-throughput screening protein purification monitoring and other biochemical applications where detergent-containing samples are analyzed
Catalog No. 50-000-97259 Supplier Apexbio Technology LLC Supplier No. K4104800
The Detergent Compatible Bradford Protein Assay Kit is designed for accurate quantification of protein concentrations in samples containing detergents Utilizing the Bradford assay principle the kit employs Coomassie Brilliant Blue dye which binds to proteins primarily through arginine and aromatic residues resulting in a measurable shift in absorbance Unlike traditional Bradford assays this formulation maintains assay performance in the presence of common detergents minimizing interference and ensuring reliable results The kit is suitable for high-throughput screening protein purification monitoring and other biochemical applications where detergent-containing samples are analyzed