Learn More
Essendant Dawn Professional Heavy Duty Liquid Degreaser, 1 gal, 3/Carton
Catalog No. 5000030071
Encompass_Preferred
Cuts through tough jobs with the grease-cutting power of Dawn This industrial degreaser is designed to clean your toughest grease problems It can be used full strength as a multi surface cleaner on buildup on range hoods fryers ovens and walls It is safe for use on food-contact surfaces when followed by rinsing with water Excellent for cleaning corral areas and dumpsters and can even be used for cleaning heavy crank case oil from concrete drive-thru pads and sidewalks This Dawn Professional product is phosphate-free noncaustic nonflammable and formulated for employee safety It s also designed to clean waxed tile floors without damaging the wax when properly diluted and can spot-clean where heavy grease buildup exists Application- Degreaser/Cleaner Applicable Material- Ceramic Glass Metal Plastic Stone Dirt Types- Carbon Grease Grime Oils Organic Matter Soil Scent- Pine
Catalog No. 50-000-30071 Supplier Essendant Supplier No. PGC75326
Cuts through tough jobs with the grease-cutting power of Dawn This industrial degreaser is designed to clean your toughest grease problems It can be used full strength as a multi surface cleaner on buildup on range hoods fryers ovens and walls It is safe for use on food-contact surfaces when followed by rinsing with water Excellent for cleaning corral areas and dumpsters and can even be used for cleaning heavy crank case oil from concrete drive-thru pads and sidewalks This Dawn Professional product is phosphate-free noncaustic nonflammable and formulated for employee safety It s also designed to clean waxed tile floors without damaging the wax when properly diluted and can spot-clean where heavy grease buildup exists Application- Degreaser/Cleaner Applicable Material- Ceramic Glass Metal Plastic Stone Dirt Types- Carbon Grease Grime Oils Organic Matter Soil Scent- Pine