Learn More
MSC Rite-Kem Detergent Disinfectant/Cleaner, Neutral, Concentrated, EPA Registered, 55 Gallon Drum
Catalog No. 502836898
Encompass
This neutral detergent disinfectant/cleaner is an EPA registered multi-purpose disinfectant, concentrated and highly dilutable. It acts as a bactericide, fungicide, and viricide, suitable for various cleaning and sanitizing needs.
  • Neutral detergent disinfectant and cleaner
  • EPA registered multi-purpose disinfectant
  • Concentrated and highly dilutable (2 oz per gallon)
  • Acts as a bactericide, fungicide, and viricide
  • Non-acid formula
  • Features a pleasant scent
  • Supplied in a 55 gallon drum
Catalog No. 50-283-6898 Supplier MSC Supplier No. 48175590
This neutral detergent disinfectant/cleaner is an EPA registered multi-purpose disinfectant, concentrated and highly dilutable. It acts as a bactericide, fungicide, and viricide, suitable for various cleaning and sanitizing needs.
  • Neutral detergent disinfectant and cleaner
  • EPA registered multi-purpose disinfectant
  • Concentrated and highly dilutable (2 oz per gallon)
  • Acts as a bactericide, fungicide, and viricide
  • Non-acid formula
  • Features a pleasant scent
  • Supplied in a 55 gallon drum