Learn More
Med Vet International Enzymatic Instrument Cleaner, 1 Gallon
Catalog No. 502831349
Encompass_Preferred
Enzymatic Detergent is a specially formulated presoak and cleaner designed to provide one step cleaning of delicate fiber optic equipment such as endoscopes and cytoscopes, as well as other surgical instruments including those for eye, cardiovascular and general surgery. It is designed as a soak solution for surgical instruments to be used immediately following their use in a surgical procedure. Hemolytic Neutral pH Bacteriostatic Double Enzyme Non-corrosive Free rinsing safe for scopes Can be used in ultrasonic cleaners Cleans hard-to-reach surfaces One gallon bottle
Catalog No. 50-283-1349 Supplier Med Vet International Supplier No. B9
Enzymatic Detergent is a specially formulated presoak and cleaner designed to provide one step cleaning of delicate fiber optic equipment such as endoscopes and cytoscopes, as well as other surgical instruments including those for eye, cardiovascular and general surgery. It is designed as a soak solution for surgical instruments to be used immediately following their use in a surgical procedure. Hemolytic Neutral pH Bacteriostatic Double Enzyme Non-corrosive Free rinsing safe for scopes Can be used in ultrasonic cleaners Cleans hard-to-reach surfaces One gallon bottle