Learn More
Hach Company Detergent, Liqui-Nox, 3.78 L (1 gallon)
Catalog No. 501995291 Shop All Hach Company Products
Encompass
Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water. Dilute 1:100. pH 8.5 Phosphate free.
Catalog No. 50-199-5291 Supplier Hach Company Supplier No. 2088117
Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water. Dilute 1:100. pH 8.5 Phosphate free.