Learn More
Hach Company Detergent, Liqui-Nox, 946 mL (1 qt)
Supplier:  Hach Company 2088153
Encompass
Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water. Dilute 1:100. pH 8.5 Phosphate free.
Catalog No. 50-199-5292