Learn More
Electron Microscopy Sciences Detergent 8 Low Foaming Phosphate Free Detergent 1 GL
SDP
Catalog No. 50997514
Encompass_Preferred
Non-ionic, concentrated detergent for labware washers, PC board washers, power-spray systems, ultrasonic cleaning systems and manual cleaning. Specially formulated to remove oils, resin rosins and fluxes from printed circuit board, electronic part, phosphate sensitive labware and nuclear contaminated equipment with no interfering conductive residues. Excellent replacement for solvent cleaning. Dilute 3:100 and it has a pH 11.
Catalog No. 50-997-514 Supplier Electron Microscopy Sciences Supplier No. 6053804
Non-ionic, concentrated detergent for labware washers, PC board washers, power-spray systems, ultrasonic cleaning systems and manual cleaning. Specially formulated to remove oils, resin rosins and fluxes from printed circuit board, electronic part, phosphate sensitive labware and nuclear contaminated equipment with no interfering conductive residues. Excellent replacement for solvent cleaning. Dilute 3:100 and it has a pH 11.