Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Detergents Test Kit
SDPSCI_ED

Catalog No. S72341 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
60 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72341 60 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S72341 Supplier LaMotte™ Supplier No. 450702
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Excellent environmental education tool for testing the properties of water

  • Test Method: Drop Count
  • Range/Sensitivity: 1 drop = 1.0 ppm Detergent
  • Number of Tests: 60 at 5.0ppm
  • Number of Reagents Used Per Test: Three
  • Anionic surfactants are extracted with toluene and break up an ion pair, releasing bromphenol blue into a water layer. Using the drop count method, a standard color reagent is then used to determine the concentration.

Specifications

For Use With (Application) Anionic surfactants are extracted with toluene and break up an ion pair, releasing bromophenol blue into a water layer, using the drop count method, a standard color reagent is then used to determine the concentration
Quantity 60 Tests
Range 1 ppm
Test Method Drop count
Type Detergent Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.