Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science Determination of Dissolved Carbon Dioxide Concentration
GREENER_CHOICESCI_ED

Catalog No. S139195 Shop All Aldon Products
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S139195 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139195 Supplier Innovating Science Supplier No. IS9757
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The proper amount of dissolved carbon dioxide in water is critical for maintaining a healthy aquatic ecosystem. Use up to 40 tests to determine dissolved carbon dioxide concentration from a water source.

  • All aquatic life has an impact on and is in turn impacted by the dissolved carbon dioxide concentration in a system, so tracking the concentration and changes in concentration can help scientists to better understand the health of an aquatic system and the organisms that exist within it.

Specifications

Grade 5+
Includes Instructional manual and all necessary chemicals
Class Size 40 Students
Product Type Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.