Learn More
Apexbio Technology LLC Detomidine HCl 90038-01-0 10mg
SDP
Catalog No. 5000092338
Encompass_Preferred
Detomidine hydrochloride (CAS 90038-01-0) is a synthetic non-narcotic small molecule that functions as an agonist at the 2-adrenergic receptor By activating these presynaptic receptors detomidine hydrochloride inhibits norepinephrine release resulting in dose-dependent sedative and analgesic effects This compound is widely utilized in biomedical research to investigate 2-adrenergic signaling pathways study mechanisms of sedation and analgesia and model neuromodulatory processes involving noradrenergic neurotransmission Its pharmacological profile makes it valuable for exploring receptor-mediated modulation within both in vitro and in vivo experimental contexts
Catalog No. 50-000-92338 Supplier Apexbio Technology LLC Supplier No. B135610
Detomidine hydrochloride (CAS 90038-01-0) is a synthetic non-narcotic small molecule that functions as an agonist at the 2-adrenergic receptor By activating these presynaptic receptors detomidine hydrochloride inhibits norepinephrine release resulting in dose-dependent sedative and analgesic effects This compound is widely utilized in biomedical research to investigate 2-adrenergic signaling pathways study mechanisms of sedation and analgesia and model neuromodulatory processes involving noradrenergic neurotransmission Its pharmacological profile makes it valuable for exploring receptor-mediated modulation within both in vitro and in vivo experimental contexts