Learn More
Medchemexpress LLC HY-B0163A 50mg Medchemexpress, Detomidine (hydrochloride) CAS:90038-01-0 Purity:>98%
SDP
Catalog No. 502082388
Encompass_Preferred
Medchemexpress, HY-B0163A 50mg Detomidine (hydrochloride) CAS:90038-01-0 Detomidine hydrochloride, an imidazole derivative, is a potent α2-adrenergic agonist. Detomidine hydrochloride produces dose-dependent sedative and analgesic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2388 Supplier Medchemexpress LLC Supplier No. HYB0163A50MG
Medchemexpress, HY-B0163A 50mg Detomidine (hydrochloride) CAS:90038-01-0 Detomidine hydrochloride, an imidazole derivative, is a potent α2-adrenergic agonist. Detomidine hydrochloride produces dose-dependent sedative and analgesic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.