Learn More
Medchemexpress LLC Detomidine hydrochloride | 90038-01-0 | 99.8% | 5 G
SDP
Catalog No. 5000406261
Encompass_Preferred
Detomidine hydrochloride is an imidazole derivative and a potent α2-adrenergic agonist. It provides dose-dependent analgesic effects through the activation of α2 catecholamine receptors, leading to reduced production of excitatory neurotransmitters. This product also has cardiac, respiratory, and antidiuretic actions due to its inhibition of the sympathetic nervous system. It is currently licensed for use in horses.
  • Potent α2-adrenergic agonist
  • Provides dose-dependent analgesic effects
  • Activates α2 catecholamine receptors
  • Inhibits the sympathetic nervous system
  • Exhibits cardiac, respiratory, and antidiuretic actions
  • Licensed for use in horses
Catalog No. 50-004-06261 Supplier Medchemexpress LLC Supplier No. HYB0163A5G
Detomidine hydrochloride is an imidazole derivative and a potent α2-adrenergic agonist. It provides dose-dependent analgesic effects through the activation of α2 catecholamine receptors, leading to reduced production of excitatory neurotransmitters. This product also has cardiac, respiratory, and antidiuretic actions due to its inhibition of the sympathetic nervous system. It is currently licensed for use in horses.
  • Potent α2-adrenergic agonist
  • Provides dose-dependent analgesic effects
  • Activates α2 catecholamine receptors
  • Inhibits the sympathetic nervous system
  • Exhibits cardiac, respiratory, and antidiuretic actions
  • Licensed for use in horses