Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Detomidine hydrochloride | 90038-01-0 | MFCD17292784 | 99.8% | 222.71 | C12H15ClN2 | 10 MG
SDP

Catalog No. 5000297389
Encompass_Preferred

Detomidine hydrochloride is the hydrochloride salt of detomidine, an imidazole-derived α2-adrenergic agonist used in research and veterinary pharmacology. It functions as a potent, dose-dependent sedative and analgesic, and is provided as a high-purity solid for in vitro and in vivo studies.

  • Potent α2-adrenergic agonist with dose-dependent sedative and analgesic effects.
  • High purity: 99.83% (as reported by supplier).
  • Molecular formula C12H15ClN2; molecular weight 222.71 g/mol.
  • Supplied as a solid in small pack sizes suitable for research (e.g., 10 MG).
  • Soluble in DMSO and water, enabling preparation of stock solutions for assays.

Catalog No. 50-002-97389 Supplier Medchemexpress LLC Supplier No. HYB0163A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detomidine hydrochloride is the hydrochloride salt of detomidine, an imidazole-derived α2-adrenergic agonist used in research and veterinary pharmacology. It functions as a potent, dose-dependent sedative and analgesic, and is provided as a high-purity solid for in vitro and in vivo studies.

  • Potent α2-adrenergic agonist with dose-dependent sedative and analgesic effects.
  • High purity: 99.83% (as reported by supplier).
  • Molecular formula C12H15ClN2; molecular weight 222.71 g/mol.
  • Supplied as a solid in small pack sizes suitable for research (e.g., 10 MG).
  • Soluble in DMSO and water, enabling preparation of stock solutions for assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.