Learn More
Cayman Chemical Detomidine (hydrochloride), Purity: ≥98%, CAS: 90038-01-0, 100 mg
SDP
Catalog No. 502966831
Encompass_Preferred
A selective α2-adrenoceptor agonist (Ki = 1.62 nM) that binds to α1-adrenoceptors with much weaker potency (Ki = 415 nM); primarily used as a sedative for horses in large animal veterinary medicine
Catalog No. 50-296-6831 Supplier Cayman Chemical Supplier No. 17853100 MG
A selective α2-adrenoceptor agonist (Ki = 1.62 nM) that binds to α1-adrenoceptors with much weaker potency (Ki = 415 nM); primarily used as a sedative for horses in large animal veterinary medicine