Learn More
Sigma Aldrich Fine Chemicals Biosciences Solujet Low Foaming Phosphate-Free Liquid Detergent | CAS No: N/A | 3.8 L
Catalog No. NC2206193
Encompass
Solujet Detergent, Low Foaming, Phosphate-Free Liquid | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 3.8LThis phosphate-free liquid detergent is designed for laboratory use. The low foaming formula can be diluted with water and is effective at removing organic residues from laboratory glassware and equipment. It is biodegradable and non-corrosive to most metals, making it safe for use on plastics, rubber and glass.
This item is not returnable. View return policy
Catalog No. NC2206193 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7429283.8L
Solujet Detergent, Low Foaming, Phosphate-Free Liquid | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 3.8LThis phosphate-free liquid detergent is designed for laboratory use. The low foaming formula can be diluted with water and is effective at removing organic residues from laboratory glassware and equipment. It is biodegradable and non-corrosive to most metals, making it safe for use on plastics, rubber and glass.