Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Detoxi-Gel™ Endotoxin Removing Columns, 1 mL

Catalog No. PI20344 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20344 5 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20344 Supplier Thermo Scientific™ Supplier No. 20344
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bind and extract endotoxins (lipopolysaccharides or LPS) from antibody or protein samples using bulk resin or columns of immobilized Polymixin B ligand.

Thermo Scientific™ Detoxi-Gel Endotoxin Removing Gel and Columns provide a quick method to remove endotoxins (lipopolysaccharides or LPS) from samples.

Detoxi-Gel Resin is a beaded agarose support onto which polymixin B has been immobilized. This ligand binds to the lipid A portion of lipopolysaccharides (LPS), which are the dominant form of endotoxins that affect biological samples. Even low levels of endotoxins can be toxic to cells or organisms and must be removed before biological samples can be added to cells for various treatment experiments. Detoxi-Gel Resin provides a quick method for removal of endotoxins from samples. It is available in convenient pre-packed columns and two sizes of slurry for use in small-scale or large-scale processes.

Highlights:

  • Affinity-based – uses immobilized polymixin B to remove endotoxins by binding to their lipid A domains

  • High capacity – 1mL of Detoxi-Gel Resin removes at least 9995 endotoxin units from a 5mL sample containing 10,000 EU of lipopolysaccharide (LPS)

  • Reusable – no loss in binding capacity even after 10 regenerations; requires activation and regeneration with 1% sodium deoxycholate (Part No. 89904)

  • Testing required – protein recovery is dependent upon protein type and concentration; empirical testing is required to validate the method for particular protein samples

  • Versatile – remove endotoxin from protein solutions, cell culture media and pharmacological components

Requires:

Sodium deoxycholate (Part No. 89904) for resin activation and regeneration.

TRUSTED_SUSTAINABILITY

Specifications

Description Detoxi-Gel Endotoxin Removing Columns, 1mL
Content And Storage Store at 4°C.
Form Liquid
Product Line Detoxi-Gel
Quantity 5 Columns
Target Endotoxin Removal
Type Endotoxin Removing Column
Stationary Phase Polymixin B
Format Spin Column
Column Type Affinity, Agarose Resin
Capacity (Metric) 1 mL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.