Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Detritylation Reagent, Solstice

Catalog No. 60090050 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
4 x 2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-050 4 x 2.5 L
60-090-208 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-050 Supplier Solstice Supplier No. BR6744X2.5GL
Add to Cart
Add to Cart

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 3% Dichloroacetic Acid in Toluene (v/v)

Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis

  • BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY

Specifications

CAS 79-43-6, 108-88-3
CAS Max % 3.00%, 97.00%
Quantity 4 x 2.5 L
Flash Point 4°C.Toluene
Packaging GL45 Glass Bottle
Physical Form Liquid, clear
Purity Grade Notes Deblocking reagents
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.