Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jade Scientific, Inc DETRITYLATION SOLUTION 20L
SDP

Catalog No. 50326198 Shop All Jade Scientific, Inc Products
Encompass_Preferred

Detritylation Solution 20l. 108-88-3 MFCD00000813

Catalog No. 50-326-198 Supplier Jade Scientific, Inc Supplier No. BJBR67420
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detritylation Solution 20l. 108-88-3 MFCD00000813

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.