Learn More
ATCC Detroit 551; Skin Fibroblast; Human (Homo sapiens)
Supplier:  ATCC CCL110
Encompass
Detroit 551 is a fibroblast cell line that was isolated in 1965 from the skin of a normal White, female embryo. This cell line was deposited by CS Stulberg.
Catalog No. 50-238-3239
 Restricted Item