Learn More
Selleck Chemical LLC Deucravacitinib S8879-25MG
SDP
Catalog No. 5000025165
Encompass_Preferred
Deucravacitinib (BMS-986165) is a highly potent and selective allosteric inhibitor of Tyk2 with a Ki value of 0 02 nM for binding to the Tyk2 pseudokinase domain It is highly selective against a panel of 265 kinases and pseudokinases
Catalog No. 50-000-25165 Supplier Selleck Chemical LLC Supplier No. S887925MG
Deucravacitinib (BMS-986165) is a highly potent and selective allosteric inhibitor of Tyk2 with a Ki value of 0 02 nM for binding to the Tyk2 pseudokinase domain It is highly selective against a panel of 265 kinases and pseudokinases