Learn More
Medchemexpress LLC Deucravacitinib | 1609392-27-9 | 99.9% | 425.46 | 200 MG
SDP
Catalog No. 5000366080
Encompass_Preferred
Deucravacitinib is a highly selective, orally bioavailable allosteric TYK2 inhibitor designed for the treatment of autoimmune diseases. It works by selectively binding to the TYK2 pseudokinase (JH2) domain, which blocks receptor-mediated Tyk2 activation by stabilizing its regulatory JH2 domain. This action inhibits IL-12/23 and type I IFN pathways. It is used for studying moderate to severe plaque psoriasis.
  • Highly selective, orally bioavailable allosteric TYK2 inhibitor
  • Designed for the treatment of autoimmune diseases
  • Selectively binds to the TYK2 pseudokinase (JH2) domain
  • Blocks receptor-mediated Tyk2 activation
  • Inhibits IL-12/23 and type I IFN pathways
  • Used for studying moderate to severe plaque psoriasis
  • FDA's first de novo deuterium
Catalog No. 50-003-66080 Supplier Medchemexpress LLC Supplier No. 200MGHY117287
Deucravacitinib is a highly selective, orally bioavailable allosteric TYK2 inhibitor designed for the treatment of autoimmune diseases. It works by selectively binding to the TYK2 pseudokinase (JH2) domain, which blocks receptor-mediated Tyk2 activation by stabilizing its regulatory JH2 domain. This action inhibits IL-12/23 and type I IFN pathways. It is used for studying moderate to severe plaque psoriasis.
  • Highly selective, orally bioavailable allosteric TYK2 inhibitor
  • Designed for the treatment of autoimmune diseases
  • Selectively binds to the TYK2 pseudokinase (JH2) domain
  • Blocks receptor-mediated Tyk2 activation
  • Inhibits IL-12/23 and type I IFN pathways
  • Used for studying moderate to severe plaque psoriasis
  • FDA's first de novo deuterium