Learn More
Azzota Corp Deut Lamp Agilent AA
Catalog No. 5000887123
Encompass_Preferred
New Deuterium UV lamp for Agilent AA instruments Replacement source lamp for background corrector used in current and most previous AA instrument models
Catalog No. 50-008-87123 Supplier Azzota Corp Supplier No. D2G8431
New Deuterium UV lamp for Agilent AA instruments Replacement source lamp for background corrector used in current and most previous AA instrument models