Learn More
Azzota Corp Deut Lamp Waters 996 2996 WAT0
Catalog No. 5000886912
Encompass_Preferred
New Deuterium Lamp for Waters 996 2996 Detectors Life time 2000 hours Compatible Manufacture part number WAT052586
Catalog No. 50-008-86912 Supplier Azzota Corp Supplier No. D2WA404
New Deuterium Lamp for Waters 996 2996 Detectors Life time 2000 hours Compatible Manufacture part number WAT052586