Learn More
Medchemexpress LLC Deutenzalutamide (enzalutamide-d3) | 1443331-82-5 | 99.8% | C21H13D3F4N4O2S | 1 MG
SDP
Catalog No. 5000433671
Encompass_Preferred
Deutenzalutamide (Enzalutamide-d3) is a deuterium-labeled form of Enzalutamide, an androgen receptor (AR) antagonist. It serves as a tracer and internal standard for quantitative analysis methods including NMR, GC-MS, or LC-MS. Deuterated compounds offer advantages such as extending in vivo half-life, improving oral bioavailability, and enhancing metabolic characteristics for drug safety.
  • Androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells
  • Functions as a tracer for quantitative analysis
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may extend in vivo half-life, improve oral bioavailability, and enhance metabolic characteristics
  • Potential to improve drug safety while retaining therapeutic properties
  • Utilized in cancer, cancer targeted therapy, and cancer metabolism research
Catalog No. 50-004-33671 Supplier Medchemexpress LLC Supplier No. HY70002S1MG
Deutenzalutamide (Enzalutamide-d3) is a deuterium-labeled form of Enzalutamide, an androgen receptor (AR) antagonist. It serves as a tracer and internal standard for quantitative analysis methods including NMR, GC-MS, or LC-MS. Deuterated compounds offer advantages such as extending in vivo half-life, improving oral bioavailability, and enhancing metabolic characteristics for drug safety.
  • Androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells
  • Functions as a tracer for quantitative analysis
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may extend in vivo half-life, improve oral bioavailability, and enhance metabolic characteristics
  • Potential to improve drug safety while retaining therapeutic properties
  • Utilized in cancer, cancer targeted therapy, and cancer metabolism research