Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium chloride 20% soln. in D2O, deuteration. deg. min. 99.5%, for NMR, MagniSolv™, MilliporeSigma™

Catalog No. M8150160025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8150160025 25 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M8150160025 Supplier MilliporeSigma Supplier No. 8150160025
Add to Cart
Add to Cart

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-07-9
Molecular Formula Cl3Ti
Molecular Weight (g/mol) 154.22
MDL Number MFCD00011266
InChI Key YONPGGFAJWQGJC-UHFFFAOYSA-K
IUPAC Name titanium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ti+3]

Specifications

Physical Form Liquid
Quantity 25 mL
Packaging Glass Bottle
Assay Percent Range 99.5% min.
Chemical Name or Material Deuterium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.