Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium chloride, 20% w/w in D{2}O, 99.5% (Isotopic)

Catalog No. AA4240718
Change view
Click to view available options
Quantity:
50 g
Packaging:
Clear Glass Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA4240718 50 g Clear Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4240718 Supplier Thermo Scientific Chemicals Supplier No. 04240718
Add to Cart

CAS: 7698-05-7 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7698-05-7
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011266
InChI Key VEXZGXHMUGYJMC-FTGQXOHASA-N
Synonym Hydrogen chloride, deuterated
IUPAC Name proton chloride
SMILES [1H+].[Cl-]

Specifications

Physical Form Liquid
Color Colorless
Packaging Clear Glass Ampule
Quantity 50 g
Sensitivity Moisture
Refractive Index 1.373
Density 1.2 g/cm3 at 20°C
Odor Acidic
Boiling Point 108°C
Assay Percent Range 99.5% (Isotopic)
Solubility Information Fully miscible in water.
Formula Weight 37.47
Chemical Name or Material Deuterium chloride, 20% w/w in D2O
Show More Show Less
Hazard Category H290-H314-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 231-715-8
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.