Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium chloride, 20% w/w in D{2}O, 99.96% (Isotopic), Thermo Scientific Chemicals

Catalog No. AA4240606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4240606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4240606 Supplier Thermo Scientific Chemicals Supplier No. 04240606
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7698-05-7 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7698-05-7
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00044642
InChI Key VEXZGXHMUGYJMC-FTGQXOHASA-N
Synonym deuterium chloride, hydrochloric acid-d, deuterium chloride, 20 wt% in deuterium oxide, chlorohydrogen, hydrochloric acid d, chloro 2h, deuterium chloride, 99 atom % d, deuterium chloride, 1 m in deuterium oxide, deuterium chloride, 0.1 m in deuterium oxide, deuterium chloride, 38 wt% in deuterium oxide
PubChem CID 522634
SMILES Cl

Specifications

Physical Form Liquid
Color Colorless to Yellow
Sensitivity Light and Moisture
Packaging Clear Glass Ampule
Quantity 5 g
Density 1.20 g/mL
Refractive Index 1.373
Odor Acidic
Boiling Point 108°C
UN Number UN1789
Solubility Information Fully miscible in water.
Formula Weight 37.47
Percent Purity ≥99.96%
Concentration or Composition (by Analyte or Components) 20% w/w in D2O
Chemical Name or Material Deuterium chloride, Isotopic
Show More Show Less
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
EINECSNumber 231-715-8
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.