Learn More
Cambridge Isotope Laboratories Deuterium Chloride (d, 99.5%) Dcl 35% W/w Solution In D2O, 7698-05-7, MFCD00044642, 50 g of solution
Catalog No. NC9382690
Encompass_Preferred
Chemical Formula: DCl, MW: 37.47, 99.5%.
Catalog No. NC9382690 Supplier Cambridge Isotope Laboratories Supplier No. DLM350
Chemical Formula: DCl, MW: 37.47, 99.5%.