Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99.5 atom % D

Catalog No. AC464140250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464140250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC464140250 Supplier Thermo Scientific Chemicals Supplier No. 464140250
Add to Cart

CAS: 7698-05-7 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7698-05-7
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
InChI Key VEXZGXHMUGYJMC-FTGQXOHASA-N
IUPAC Name proton chloride
SMILES [1H+].[Cl-]

Specifications

Refractive Index 1.3726
Specific Gravity 1.2
Quantity 25 mL
Density 1.2000 g/mL
Boiling Point 108.0°C
Linear Formula DCl
Formula Weight 37.47
Grade NMR
Chemical Name or Material Deuterium chloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May be corrosive to metals.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF exposed or concerned: Get medical advice/attention.

If skin irritation occurs: Get medical advice/attention.

If eye irritation persists: Get medical advice/attention.

EINECSNumber 231-715-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.