Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D

Catalog No. AC175420500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175420500 50 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175420500 Supplier Thermo Scientific Chemicals Supplier No. 175420500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7698-05-7 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7698-05-7
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00044642
InChI Key VEXZGXHMUGYJMC-FTGQXOHASA-N
Synonym deuterium chloride, hydrochloric acid-d, deuterium chloride, 20 wt% in deuterium oxide, chlorohydrogen, hydrochloric acid d, chloro 2h, deuterium chloride, 99 atom % d, deuterium chloride, 1 m in deuterium oxide, deuterium chloride, 0.1 m in deuterium oxide, deuterium chloride, 38 wt% in deuterium oxide
PubChem CID 522634
SMILES Cl

Specifications

Physical Form Liquid
Color Colorless
Quantity 50 g
Density 1.2000g/mL
Packaging Glass Bottle
Refractive Index 1.3726
Deuterium (D) 99.0% min.
Boiling Point 108.0°C
Linear Formula DCl
Formula Weight 37.47
Grade NMR
Chemical Name or Material Deuterium chloride, 99+ atom % D
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.