Learn More
Cambridge Isotope Laboratories DEUTERIUM (D, 99.9%) (D2,99.8%+HD,0.2%), 850 L, 7782-39-0
Catalog No. 501630874
Encompass
DEUTERIUM (D, 99.9%) (D2,99.8%+HD,0.2%), 850 L, 7782-39-0 **This item is shipped in a CODE H-8, 8L CARBON STEEL CYLINDER. The fee for the cylinder will need added to the order as a separate charge, per each. Please include part# NC2038865 to your order.
Catalog No. 50-163-0874 Supplier Cambridge Isotope Laboratories Supplier No. DLM1329850
DEUTERIUM (D, 99.9%) (D2,99.8%+HD,0.2%), 850 L, 7782-39-0 **This item is shipped in a CODE H-8, 8L CARBON STEEL CYLINDER. The fee for the cylinder will need added to the order as a separate charge, per each. Please include part# NC2038865 to your order.