Learn More
World Precision Instrument Deuterium/Halogen Fiber Light Source
SDP
Catalog No. 502538872
Encompass
For a continuous spectrum in the UV, VIS and NIR range
The D4H is a combined deuterium and halogen light source for UV/VIS and NIR applications. This light source is ideally suited to work with WPI’s spectrometer modules and sample cells. It supplies a continuous spectrum in the UV, VIS and NIR range from 200 nm to 1700 nm. The D4H is equipped with an integrated electrical shutter, which can be controlled by a switch or a TTL signal.
Catalog No. 50-253-8872 Supplier World Precision Instrument Supplier No. D4H
Add to Cart
Add to Cart
Request a Quote
For a continuous spectrum in the UV, VIS and NIR range
The D4H is a combined deuterium and halogen light source for UV/VIS and NIR applications. This light source is ideally suited to work with WPI’s spectrometer modules and sample cells. It supplies a continuous spectrum in the UV, VIS and NIR range from 200 nm to 1700 nm. The D4H is equipped with an integrated electrical shutter, which can be controlled by a switch or a TTL signal.