Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deuterium labeled MCPA | 352431-14-2 | 98.0% | 203.64 | C9H6D3ClO3 | 10 MG
SDP

Catalog No. 5000296743
Encompass_Preferred

MCPA-d3 is the deuterium-labeled analog of the phenoxyacetic acid herbicide MCPA, provided as a solid for research applications. It is used as a tracer or internal standard for quantitative analysis and for studies of herbicide action and metabolism.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • Off-white solid provided in a 10 mg quantity.
  • Purity ≥98.0% for analytical use.
  • CAS number 352431-14-2 to aid substance identification.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-96743 Supplier Medchemexpress LLC Supplier No. HYB0859S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MCPA-d3 is the deuterium-labeled analog of the phenoxyacetic acid herbicide MCPA, provided as a solid for research applications. It is used as a tracer or internal standard for quantitative analysis and for studies of herbicide action and metabolism.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • Off-white solid provided in a 10 mg quantity.
  • Purity ≥98.0% for analytical use.
  • CAS number 352431-14-2 to aid substance identification.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.