Learn More
Medchemexpress LLC Deuterium labeled uridine | 40632-21-1 | 98.5% | 10 MG

Supplier: Medchemexpress LLC HYB1449S810MG
Deuterium-labeled uridine supplied as a stable-isotope standard for analytical and biochemical research. The compound is intended for use as an internal standard, tracer, or reference in LC-MS quantification, metabolite identification, and metabolic flux studies. It is provided at high reported purity to support reproducible analytical results.
- Molecular formula C9H10D2N2O6
- Molecular weight 246.21 g/mol
- Purity 98.5%
- CAS number 40632-21-1
- Intended use LC-MS, metabolite identification, and tracer studies
- Available in small milligram quantities for analytical workflows
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.