Learn More
Phenomenex Inc Deuterium lamp-2000h
Catalog No. 502394227 Shop All Phenomenex Products
Encompass_Preferred
Deuterium lamp-2000h with double hole; used with tungsten lamp
Catalog No. 50-239-4227 Supplier Phenomenex Inc Supplier No. 14000008
Deuterium lamp-2000h with double hole; used with tungsten lamp