Learn More
Azzota Corp Deuterium Lamp Agilent
Catalog No. 5000023480
Encompass_Preferred
Deuterium Lamp Equivalent to Agilent PN 08452-60104 for Agilent 8452A
Catalog No. 50-000-23480 Supplier Azzota Corp Supplier No. D28452A
Deuterium Lamp Equivalent to Agilent PN 08452-60104 for Agilent 8452A