Learn More
Optimize Technologies Deuterium Lamp, Dionex CES I, VDM-2
SDP
Catalog No. 50113702
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. 
Catalog No. 50-113-702 Supplier Optimize Technologies Supplier No. 253202820
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. 