Learn More
Perkin Elmer US LLC Deuterium Lamp for PinAAcle 500/900 AA
Catalog No. 502104891
Encompass_Preferred
This Deuterium (D2) lamp is used for background correction in flame AA instruments such as the PinAAcle 500 and 900F, as well as combination flame/furnace instruments such as the PinAAcle 900H and 900T.
Catalog No. 50-210-4891 Supplier Perkin Elmer US LLC Supplier No. N3160811
This Deuterium (D2) lamp is used for background correction in flame AA instruments such as the PinAAcle 500 and 900F, as well as combination flame/furnace instruments such as the PinAAcle 900H and 900T.