Learn More
Optimize Technologies Deuterium Lamp, Spectra Physics Focus, 100, 200, UV-100, -150, -200, -100, 2000, 3000, Focus, CE Series
SDP
Catalog No. 50113999
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is designed to work with various Spectra Physics pump models and is equivalent to OEM part number 9551-0023.
Catalog No. 50-113-999 Supplier Optimize Technologies Supplier No. 353201333
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is designed to work with various Spectra Physics pump models and is equivalent to OEM part number 9551-0023.