Learn More
Agilent Technologies Long-Life Deuterium VWD Lamp, Fused Silica, 1 ea
Catalog No. NC3712832
Encompass
The long-life deuterium lamp is designed for optimal performance in G1314A/B/C variable wavelength detectors and 1120 and 1220 Infinity LC systems. Constructed from fused silica, this lamp provides high intensity across the 190 to 600 nm spectral range to ensure superior signal-to-noise ratios, sensitivity, and stability for chromatographic analysis.
  • Guaranteed lifetime of greater than 2,000 hr with minimal intensity drop
  • Narrower aperture design provides decreased noise and lower detection limits
  • Superior signal stability and shorter stabilization time after ignition
  • Safety-focused design includes a bulky cap to block UV exposure and a double-insulated cable to prevent electrical shock
  • Rigorous quality assurance testing for light intensity, noise, drift, and alignment
  • Consistent performance enabled by automated, ISO 9001-certified manufacturing processes
Catalog No. NC3712832 Supplier Agilent Technologies Supplier No. G131460100
The long-life deuterium lamp is designed for optimal performance in G1314A/B/C variable wavelength detectors and 1120 and 1220 Infinity LC systems. Constructed from fused silica, this lamp provides high intensity across the 190 to 600 nm spectral range to ensure superior signal-to-noise ratios, sensitivity, and stability for chromatographic analysis.
  • Guaranteed lifetime of greater than 2,000 hr with minimal intensity drop
  • Narrower aperture design provides decreased noise and lower detection limits
  • Superior signal stability and shorter stabilization time after ignition
  • Safety-focused design includes a bulky cap to block UV exposure and a double-insulated cable to prevent electrical shock
  • Rigorous quality assurance testing for light intensity, noise, drift, and alignment
  • Consistent performance enabled by automated, ISO 9001-certified manufacturing processes